Learn More
Ace Glass, Inc. 13mm ID 8" 24/40 epoxy coated flash chromatography column with porosity B filter disc
SDP
Catalog No. 50972435
Encompass_Preferred
Catalog No. 50-972-435 Supplier Ace Glass, Inc. Supplier No. 587151