Learn More
Ace Glass, Inc. 19mm ID 10" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972504
Encompass_Preferred
19mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-504 Supplier Ace Glass, Inc. Supplier No. 587262
19mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.