Learn More
Ace Glass, Inc. 19mm ID 10" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50978562
Encompass_Preferred
19mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-978-562 Supplier Ace Glass, Inc. Supplier No. 587208
19mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.