Learn More
Ace Glass, Inc. 19mm ID 18" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972498
Encompass_Preferred
19mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-498 Supplier Ace Glass, Inc. Supplier No. 587254
19mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.