Learn More
Ace Glass, Inc. 19mm ID 18" 24/40 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972440
Encompass_Preferred
19mm I.D. x 457mm long, simple absorption chromatography column for rapid preparative separations. Column is epoxy coated for safety, and has 70-100 micron porosity sealed-in fritted disc above the 2mm bore PTFE stopcock. 130mL capacity.
Catalog No. 50-972-440 Supplier Ace Glass, Inc. Supplier No. 587162
19mm I.D. x 457mm long, simple absorption chromatography column for rapid preparative separations. Column is epoxy coated for safety, and has 70-100 micron porosity sealed-in fritted disc above the 2mm bore PTFE stopcock. 130mL capacity.