Learn More
Ace Glass, Inc. 19mm ID 24" 24/40 epoxy coated flash chromatography column with porosity B filter disc
SDP
Catalog No. 50972567
Encompass_Preferred
19mm I.D., 173mL epoxy coated chromatography column with a 24/40 standard taper joint at the top, and a 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom of the column. Column has an effective length of 610mm, and is supplied with a porosity B (70-100 micron) fritted disc sealed near the bottom of the tube.
Catalog No. 50-972-567 Supplier Ace Glass, Inc. Supplier No. 590426
19mm I.D., 173mL epoxy coated chromatography column with a 24/40 standard taper joint at the top, and a 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom of the column. Column has an effective length of 610mm, and is supplied with a porosity B (70-100 micron) fritted disc sealed near the bottom of the tube.