Learn More
Ace Glass, Inc. 19mm ID 8" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972467
Encompass_Preferred
19mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-467 Supplier Ace Glass, Inc. Supplier No. 587207
19mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.