Learn More
Ace Glass, Inc. 25mm ID 10" 24/40 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972441
Encompass_Preferred
Catalog No. 50-972-441 Supplier Ace Glass, Inc. Supplier No. 587167