Learn More
Ace Glass, Inc. 25mm ID 12" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972507
Encompass_Preferred
25mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-507 Supplier Ace Glass, Inc. Supplier No. 587266
25mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.