Learn More
Ace Glass, Inc. 25mm ID 12" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972472
Encompass_Preferred
25mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-472 Supplier Ace Glass, Inc. Supplier No. 587215
25mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.