Learn More
Ace Glass, Inc. 25mm ID 18" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972508
Encompass_Preferred
25mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-508 Supplier Ace Glass, Inc. Supplier No. 587267
25mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.