Learn More
Ace Glass, Inc. 25mm ID 18" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972473
Encompass_Preferred
25mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-473 Supplier Ace Glass, Inc. Supplier No. 587216
25mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.