Learn More
Neta Scientific COLUMN 300A 7.8X150MM 2.7UM
SDP
Catalog No. NC2032481
Encompass_Preferred
This item is manufactured by Agilent
AdvanceBio SEC 300, 7.8 x 150 mm, 2.7 m, LC column
Catalog No. NC2032481 Supplier Neta Scientific Supplier No. PL11803301
This item is manufactured by Agilent
AdvanceBio SEC 300, 7.8 x 150 mm, 2.7 m, LC column