Learn More
Ace Glass, Inc. 38mm ID 10" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50978576
Encompass_Preferred
38mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-978-576 Supplier Ace Glass, Inc. Supplier No. 587269
38mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.