Learn More
Ace Glass, Inc. 38mm ID 12" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972510
Encompass_Preferred
38mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-510 Supplier Ace Glass, Inc. Supplier No. 587270
38mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.