Learn More
Ace Glass, Inc. 38mm ID 12" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972475
Encompass_Preferred
38mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-475 Supplier Ace Glass, Inc. Supplier No. 587219
38mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.