Learn More
Ace Glass, Inc. 38mm ID 18" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50978565
Encompass_Preferred
38mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-978-565 Supplier Ace Glass, Inc. Supplier No. 587222
38mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.