Learn More
Ace Glass, Inc. 41mm ID 18" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972500
Encompass_Preferred
41mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-500 Supplier Ace Glass, Inc. Supplier No. 587256
41mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.