Learn More
Ace Glass, Inc. 41mm ID 18" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972471
Encompass_Preferred
41mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-471 Supplier Ace Glass, Inc. Supplier No. 587214
41mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.