Learn More
Ace Glass, Inc. 50mm ID 10" 50/30 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50978577
Encompass_Preferred
50mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints. Includes porosity B filter disc.
Catalog No. 50-978-577 Supplier Ace Glass, Inc. Supplier No. 587273
50mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints. Includes porosity B filter disc.