Learn More
Ace Glass, Inc. 50mm ID 10" 50/30 epoxy coated flash chromatography column.
SDP
Catalog No. 50972478
Encompass_Preferred
50mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.
Catalog No. 50-972-478 Supplier Ace Glass, Inc. Supplier No. 587224
50mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.