Learn More
Ace Glass, Inc. 50mm ID 18" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972476
Encompass_Preferred
50mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-476 Supplier Ace Glass, Inc. Supplier No. 587220
50mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.