Learn More
Ace Glass, Inc. 50mm ID 24" 24/40 epoxy coated flash chromatography column
SDP
Catalog No. 50972565
Encompass_Preferred
50mm I.D. x 24 L epoxy coated flash chromatography column with 24/40 standard taper joint at top, and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with nylon bushing, FETFE o-ring, and tubing connector for connecting 12.5-14mm O.D. tubing to the column.
Catalog No. 50-972-565 Supplier Ace Glass, Inc. Supplier No. 590328
50mm I.D. x 24 L epoxy coated flash chromatography column with 24/40 standard taper joint at top, and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with nylon bushing, FETFE o-ring, and tubing connector for connecting 12.5-14mm O.D. tubing to the column.