Learn More
Ace Glass, Inc. 50mm ID 8" 50/30 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972512
Encompass_Preferred
50mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints. Includes porosity B filter disc.
Catalog No. 50-972-512 Supplier Ace Glass, Inc. Supplier No. 587272
50mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints. Includes porosity B filter disc.