Learn More
Ace Glass, Inc. 64mm ID 18" 50/30 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972517
Encompass_Preferred
64mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints. Includes porosity B filter disc.
Catalog No. 50-972-517 Supplier Ace Glass, Inc. Supplier No. 587282
64mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints. Includes porosity B filter disc.