Learn More
Ace Glass, Inc. 64mm ID 18" 45/50 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972452
Encompass_Preferred
Catalog No. 50-972-452 Supplier Ace Glass, Inc. Supplier No. 587190