Learn More
Ace Glass, Inc. 64mm ID 18" 50/30 epoxy coated flash chromatography column.
SDP
Catalog No. 50972483
Encompass_Preferred
64mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.
Catalog No. 50-972-483 Supplier Ace Glass, Inc. Supplier No. 587231
64mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.