Learn More
Ace Glass, Inc. 64mm ID 24" 50/30 epoxy coated flash chromatography column.
SDP
Catalog No. 50972484
Encompass_Preferred
64mm I.D., 24 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.
Catalog No. 50-972-484 Supplier Ace Glass, Inc. Supplier No. 587232
64mm I.D., 24 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.