Learn More
Ace Glass, Inc. 75mm ID 12" 75/50 epoxy coated flash chromatography column.
SDP
Catalog No. 50972496
Encompass_Preferred
75mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 75/50 joints.
Catalog No. 50-972-496 Supplier Ace Glass, Inc. Supplier No. 587251
75mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 75/50 joints.