Learn More
Ace Glass, Inc. 75mm ID 24" 75/50 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50978580
Encompass_Preferred
75mm I.D., 24 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 75/50 joints. Includes porosity B filter disc.
Catalog No. 50-978-580 Supplier Ace Glass, Inc. Supplier No. 587298
75mm I.D., 24 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 75/50 joints. Includes porosity B filter disc.