Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 1000mL Reservoir, 2 1/2in ID X 12in E.L., 4mm Stpk
SDP
Supplier:  Chemglass Life Sciences CG120333
Encompass_Preferred
Column, Chromatography, 35/20 Socket Joint, 1000mL Reservoir, 2 1/2in ID X 12in E.L., 4mm Stopcock. Similar to CG-1202 but with a COARSE porosity fritted disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with CG-1201 flow controller. Connection between the column and flow control adapter are spherical joints connected with CG-150 stainless steel clamps.
Catalog No. CG120333