Learn More
Teledyne ISCO, Inc COLUMN ALUMINA NEUTRAL 10PK
Catalog No. 50871504 Shop All Teledyne ISCO Inc. Products
Encompass_Preferred
160 gram Alumina Neutral RediSep Rf columns, pkg. of 12.
Catalog No. 50-871-504 Supplier Teledyne ISCO, Inc Supplier No. 692203446
160 gram Alumina Neutral RediSep Rf columns, pkg. of 12.