Learn More
Bio Rad Laboratories COLUMN AMINEX HPX-42A
Catalog No. 509012152
Encompass
Columns, Carbohydrate Analysis; Bio-Rad; Aminex; HPX-42A; 300 x 7.8mm; Temperature: 85 deg.C; Flow rate: 0.6mL/min
Catalog No. 50-901-2152 Supplier Bio Rad Laboratories Supplier No. 1250097
Columns, Carbohydrate Analysis; Bio-Rad; Aminex; HPX-42A; 300 x 7.8mm; Temperature: 85 deg.C; Flow rate: 0.6mL/min