Learn More
Microsolv Technology Corporation COLUMN BIDENTATE C18 HPLCCOLU
Catalog No. 5001451089
Encompass_Preferred
Cogent Bidentate C18 HPLC Column. 4um 100A 75mm x 4.6mm ID.
Catalog No. 50-014-51089 Supplier Microsolv Technology Corporation Supplier No. 4001875P
Cogent Bidentate C18 HPLC Column. 4um 100A 75mm x 4.6mm ID.