Learn More
Microsolv Technology Corporation Column Bidentate C18 Hplccolu
Supplier:  Microsolv Technology Corporation 4001815P
Encompass_Preferred
HPLC Column, Bidentate C18, 4um, 4.6mm ID x 150mm Length, 100A. Cogent TYPE-C Brand. 1/EA.
Catalog No. 50-014-51082