Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ column Blank Kit for HPLC

Catalog No. 111016267 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
I.D. O.D. L 10 mm 1/2 in. 25 cm
I.D. O.D. L 4.6 mm 1/4 in. 15 cm
I.D. O.D. L 4.6 mm 1/4 in. 25 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-101-6267 I.D. O.D. L 4.6 mm 1/4 in. 25 cm
11-101-0713 I.D. O.D. L 4.6 mm 1/4 in. 15 cm
11-101-0261 I.D. O.D. L 10 mm 1/2 in. 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-6267 Supplier MilliporeSigma™ Supelco™ Supplier No. 59102U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Each column blank kit includes a precision-bore polished 316 stainless steel column blank, two zero dead volume external reducing unions (internal unions with 10 mm I.D. blanks) with nuts and ferrules for 1/16 in. O.D. tubing, and two 2 μm frits.

TRUSTED_SUSTAINABILITY

Specifications

Material 316 L Stainless Steel
Description I.D. O.D. L 4.6 mm 1/4 in. 25 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.