Learn More
Trajan Scientific Column BP1 QC1 5 in 1/EA
SDP
Catalog No. 5000494246
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length BP1 GC capillary column on 5 cage
Catalog No. 50-004-94246 Supplier Trajan Scientific Supplier No. 054022CCS
0 1 mm ID x 0 1 m film thickness x 10 m length BP1 GC capillary column on 5 cage