Learn More
Trajan Scientific Column BP1 QC1 Medical 1/EA
SDP
Catalog No. 5000494882
Encompass_Preferred
0 1 mm ID x 0 1 m film thickness x 10 m length BP1 GC capillary column
Catalog No. 50-004-94882 Supplier Trajan Scientific Supplier No. 054022
0 1 mm ID x 0 1 m film thickness x 10 m length BP1 GC capillary column