Learn More
Trajan Scientific Column BP1 QC2.5 5 in 1/EA
SDP
Catalog No. 5000494381
Encompass_Preferred
0 25 mm ID x 1 m film thickness x 60 m length BP1 GC capillary column on 5 cage
Catalog No. 50-004-94381 Supplier Trajan Scientific Supplier No. 054815CCS
Add to Cart
Add to Cart
0 25 mm ID x 1 m film thickness x 60 m length BP1 GC capillary column on 5 cage