Learn More
Trajan Scientific Column BP1 QC2.5 Medical 1/EA
SDP
Catalog No. 5000493563
Encompass_Preferred
0 25 mm ID x 1 m film thickness x 30 m length BP1 GC capillary column
Catalog No. 50-004-93563 Supplier Trajan Scientific Supplier No. 054056
0 25 mm ID x 1 m film thickness x 30 m length BP1 GC capillary column