Learn More
Trajan Scientific Column BP1 QC2 Medical 1/EA
SDP
Catalog No. 5000494499
Encompass_Preferred
0 22 mm ID x 1 m film thickness x 50 m length BP1 GC capillary column
Catalog No. 50-004-94499 Supplier Trajan Scientific Supplier No. 054054
0 22 mm ID x 1 m film thickness x 50 m length BP1 GC capillary column