Learn More
Trajan Scientific Column BP1 QC3 Medical 1/EA
SDP
Catalog No. 5000494886
Encompass_Preferred
0 32 mm ID x 5 m film thickness x 60 m length BP1 GC capillary column
Catalog No. 50-004-94886 Supplier Trajan Scientific Supplier No. 054085
0 32 mm ID x 5 m film thickness x 60 m length BP1 GC capillary column