Learn More
Trajan Scientific Column BP1 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054065
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 25 m length BP1 GC capillary column
Catalog No. 50-004-94039