Learn More
Trajan Scientific Column BP1 QC3 Medical 1/EA
SDP
Catalog No. 5000493607
Encompass_Preferred
0 32 mm ID x 3 m film thickness x 30 m length BP1 GC capillary column
Catalog No. 50-004-93607 Supplier Trajan Scientific Supplier No. 054073
0 32 mm ID x 3 m film thickness x 30 m length BP1 GC capillary column