Learn More
Trajan Scientific Column BP1 QC5 Medical 1/EA
SDP
Catalog No. 5000493647
Encompass_Preferred
0 53 mm ID x 1 m film thickness x 30 m length BP1 GC capillary column
Catalog No. 50-004-93647 Supplier Trajan Scientific Supplier No. 054090
0 53 mm ID x 1 m film thickness x 30 m length BP1 GC capillary column