Learn More
Trajan Scientific Column BP1 QC5 Medical 1/EA
SDP
Catalog No. 5000494417
Encompass_Preferred
0 53 mm ID x 1 m film thickness x 12 m length BP1 GC capillary column
Catalog No. 50-004-94417 Supplier Trajan Scientific Supplier No. 054086
0 53 mm ID x 1 m film thickness x 12 m length BP1 GC capillary column