Learn More
Trajan Scientific Column BP10 QC2.5 Medical 1/EA
SDP
Catalog No. 5000494233
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length BP10 GC capillary column
Catalog No. 50-004-94233 Supplier Trajan Scientific Supplier No. 054256
0 25 mm ID x 0 25 m film thickness x 30 m length BP10 GC capillary column