Learn More
Trajan Scientific Column BP10 QC2 Medical 1/EA
SDP
Catalog No. 5000494232
Encompass_Preferred
0 22 mm ID x 0 25 m film thickness x 25 m length BP10 GC capillary column
Catalog No. 50-004-94232 Supplier Trajan Scientific Supplier No. 054253
0 22 mm ID x 0 25 m film thickness x 25 m length BP10 GC capillary column