Learn More
Trajan Scientific Column BP10 QC3 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054259
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length BP10 GC capillary column
Catalog No. 50-004-93974