Learn More
Trajan Scientific Column BP10 QC5 Medical 1/EA
SDP
Catalog No. 5000494884
Encompass_Preferred
0 53 mm ID x 1 0 m film thickness x 30 m length BP10 GC capillary column
Catalog No. 50-004-94884 Supplier Trajan Scientific Supplier No. 054283
0 53 mm ID x 1 0 m film thickness x 30 m length BP10 GC capillary column